Assay Details
Target Gene Details
Entrez Gene ID: | 84876 |
Gene Name: | ORAI calcium release-activated calcium modulator 1 |
Gene Aliases: |
CRACM1, IMD9, ORAT1, TAM2, TMEM142A |
Location: |
Chr.12:121626550-121642040 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ORAI1 | NM_032790.3 | 1 | 427 | NP_116179.2 |
| AK027372.1 | 1 | 171 | BAB55068.1 | |
| BC013386.1 | 1 | 331 | AAH13386.1 | |
| BC015369.1 | 1 | 289 | AAH15369.2 | |
| BC075831.1 | 1 | 427 | AAH75831.1 | |
| BC104633.1 | 1 | 192 | AAI04634.1 | |
| BG775018.1 | 1 | 154 | ||
| BM762473.1 | 1 | 351 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560458 | Chr.12:121598620 - 121715590 on Build GRCh38 | Loss |
|
| nsv1053272 | Chr.12:121507127 - 121694571 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map