Assay Details
Target Gene Details
Entrez Gene ID: | 55818 |
Gene Name: | lysine demethylase 3A |
Gene Aliases: |
JHDM2A, JHMD2A, JMJD1, JMJD1A, TSGA |
Location: |
Chr.2:86440647-86492716 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM3A | NM_001146688.1 | 11 | 1699 | NP_001140160.1 |
| NM_018433.5 | 11 | 1968 | NP_060903.2 | |
| XM_006712051.3 | 10 | 1671 | XP_006712114.1 | |
| XM_017004493.1 | 11 | 1987 | XP_016859982.1 | |
| XM_017004494.1 | 12 | 1810 | XP_016859983.1 | |
| AB018285.2 | 11 | 1731 | ||
| AK304276.1 | 9 | 1485 | ||
| AL832150.1 | 11 | 1734 | CAH18459.3 | |
| BX640698.1 | 11 | 1769 | CAE45820.1 | |
| BX647271.1 | 11 | 1737 | ||
| BX647752.1 | 11 | 1797 | ||
| BX648841.1 | 11 | 1700 | ||
| BX649109.1 | 11 | 1738 | ||
| CR749581.1 | 11 | 1736 | CAH18373.1 |
More Information
Set Membership: |
|

Genomic Map