Assay Details
Target Gene Details
Entrez Gene ID: | 5058 |
Gene Name: | p21 (RAC1) activated kinase 1 |
Gene Aliases: |
PAKalpha |
Location: |
Chr.11:77322015-77474063 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAK1 | NM_001128620.1 | 16 | 2717 | NP_001122092.1 |
| NM_002576.4 | 15 | 2668 | NP_002567.3 | |
| XM_011545080.2 | 16 | 2424 | XP_011543382.1 | |
| XM_017017846.1 | 15 | 2177 | XP_016873335.1 | |
| XM_017017847.1 | 15 | 2375 | XP_016873336.1 | |
| XM_017017848.1 | 15 | 2444 | XP_016873337.1 | |
| XM_017017849.1 | 15 | 2321 | XP_016873338.1 | |
| XM_017017850.1 | 16 | 2384 | XP_016873339.1 | |
| AK055228.1 | 15 | 2129 | ||
| BC109299.1 | 15 | 2276 | AAI09300.1 |
More Information
Set Membership: |
|

Genomic Map