Assay Details
Target Gene Details
Entrez Gene ID: | 7066 |
Gene Name: | thrombopoietin |
Gene Aliases: |
MGDF, MKCSF, ML, MPLLG, THCYT1, TPO |
Location: |
Chr.3:184371935-184379688 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THPO | NM_000460.3 | NP_000451.1 | ||
| NM_001177597.2 | NP_001171068.1 | |||
| NM_001177598.2 | NP_001171069.1 | |||
| NM_001289997.1 | NP_001276926.1 | |||
| NM_001289998.1 | NP_001276927.1 | |||
| NM_001290003.1 | NP_001276932.1 | |||
| NM_001290022.1 | NP_001276951.1 | |||
| NM_001290026.1 | NP_001276955.1 | |||
| NM_001290027.1 | NP_001276956.1 | |||
| NM_001290028.1 | NP_001276957.1 | |||
| XM_017007107.1 | XP_016862596.1 | |||
| BC130322.1 | ||||
| BC136338.1 | ||||
| BC143982.1 | ||||
| L33410.1 | AAA59857.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422400 | Chr.3:184299495 - 184425713 on Build GRCh38 | Duplication |
|
| nsv829813 | Chr.3:184264997 - 184427996 on Build GRCh38 | Loss |
|
| nsv592667 | Chr.3:184375988 - 184380341 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map