Assay Details
Target Gene Details
Entrez Gene ID: | 79573 |
Gene Name: | tetratricopeptide repeat domain 13 |
Gene Aliases: |
- |
Location: |
Chr.1:230906241-230978872 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC13 | NM_001122835.2 | 11 | 1431 | NP_001116307.2 |
| NM_024525.4 | 13 | 1590 | NP_078801.3 | |
| XM_005273260.1 | 13 | 1590 | XP_005273317.1 | |
| XM_005273261.2 | 12 | 1497 | XP_005273318.1 | |
| XM_005273262.1 | 11 | 1431 | XP_005273319.1 | |
| XM_005273264.2 | 11 | 1362 | XP_005273321.1 | |
| XM_006711814.2 | 9 | 1203 | XP_006711877.1 | |
| XM_006711815.2 | 6 | 2597 | XP_006711878.1 | |
| XM_011544275.2 | 10 | 1269 | XP_011542577.1 | |
| XM_011544276.2 | 6 | 833 | XP_011542578.1 | |
| XM_017002321.1 | 12 | 1497 | XP_016857810.1 | |
| XM_017002322.1 | 11 | 1362 | XP_016857811.1 | |
| XM_017002323.1 | 9 | 1165 | XP_016857812.1 | |
| XM_017002324.1 | 9 | 1203 | XP_016857813.1 | |
| XM_017002325.1 | 13 | 4795 | XP_016857814.1 | |
| XM_017002326.1 | 8 | 4175 | XP_016857815.1 | |
| AK074365.1 | 4 | 498 | ||
| AK075386.1 | 13 | 1556 | BAC11586.1 | |
| AK075561.1 | 11 | 1410 | BAC11700.1 | |
| AK309725.1 | 9 | 1249 | ||
| AL832286.1 | 13 | 1325 | CAH10599.1 | |
| AL834453.1 | 4 | 559 | CAD39113.1 | |
| BC042683.1 | 4 | 477 | AAH42683.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3589093 | Chr.1:230482037 - 231277190 on Build GRCh38 | Gain |
|
| nsv1000488 | Chr.1:230799658 - 231269735 on Build GRCh38 | Gain |
|

Genomic Map