Assay Details
Target Gene Details
Entrez Gene ID: | 113802 |
Gene Name: | HEN1 methyltransferase homolog 1 |
Gene Aliases: |
C1orf59, HEN1 |
Location: |
Chr.1:108648288-108661526 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HENMT1 | NM_144584.2 | 1 | 260 | NP_653185.2 |
| XM_017000179.1 | 1 | 160 | XP_016855668.1 | |
| AK055087.1 | 1 | 260 | BAB70852.1 | |
| AK310152.1 | 1 | 250 | ||
| AK311542.1 | 1 | 160 | ||
| BC012198.1 | AAH12198.1 |
More Information
Set Membership: |
|

Genomic Map