Assay Details
Target Gene Details
Entrez Gene ID: | 3381 |
Gene Name: | integrin binding sialoprotein |
Gene Aliases: |
BNSP, BSP, BSP-II, SP-II |
Location: |
Chr.4:87799550-87812449 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IBSP | NM_004967.3 | 7 | 1050 | NP_004958.2 |
| BQ573754.1 | 1 | 433 | ||
| J05213.1 | AAC95490.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv322n21 | Chr.4:87811611 - 87812379 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map