Assay Details
Target Gene Details
Entrez Gene ID: | 2997 |
Gene Name: | glycogen synthase 1 |
Gene Aliases: |
GSY, GYS |
Location: |
Chr.19:48968125-48993353 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GYS1 | NM_001161587.1 | NP_001155059.1 | ||
| NM_002103.4 | NP_002094.2 | |||
| NR_027763.1 | ||||
| AK223579.1 | BAD97299.1 | |||
| AK291672.1 | ||||
| AK294349.1 | ||||
| AK300369.1 | ||||
| AK302738.1 | ||||
| AK303712.1 | ||||
| BC002617.2 | AAH02617.1 | |||
| BC003182.1 | AAH03182.1 | |||
| BC007688.2 | AAH07688.1 | |||
| J04501.1 | AAA88046.1 | |||
| U32573.1 | AAB60385.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| dgv3618n100 | Chr.19:48948950 - 49021211 on Build GRCh38 | Gain |
|
| nsv833857 | Chr.19:48914476 - 49146320 on Build GRCh38 | Loss |
|
| nsv953598 | Chr.19:48957644 - 49085143 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map