Assay Details
Target Gene Details
Entrez Gene ID: | 54984 |
Gene Name: | PIN2/TERF1 interacting, telomerase inhibitor 1 |
Gene Aliases: |
LPTL, LPTS |
Location: |
Chr.8:10764961-10839899 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PINX1 | NM_001284356.1 | 6 | 976 | NP_001271285.1 |
| NM_017884.5 | 7 | 1053 | NP_060354.4 | |
| AF205718.1 | 6 | 965 | AAG18009.1 | |
| AF418553.1 | 7 | 1042 | AAN31333.1 | |
| AJ344104.1 | 7 | 1042 | CAC51436.1 | |
| AK313715.1 | 7 | 991 | ||
| AY238941.1 | 6 | 858 | AAP37006.1 | |
| AY523566.1 | 7 | 935 | AAS19507.1 | |
| BC015479.1 | 7 | 999 | AAH15479.1 | |
| BC093762.1 | 7 | 943 | AAH93762.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7012n100 | Chr.8:10747928 - 10805045 on Build GRCh38 | Loss |
|
| nsv831232 | Chr.8:10748543 - 10917438 on Build GRCh38 | Loss |
|
| dgv7011n100 | Chr.8:10734291 - 10946733 on Build GRCh38 | Gain |
|
| esv2764105 | Chr.8:10753047 - 10800216 on Build GRCh38 | Loss |
|
| dgv447n21 | Chr.8:10742744 - 10834483 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map