Assay Details
Target Gene Details
Entrez Gene ID: | 55110 |
Gene Name: | mago homolog B, exon junction complex core component |
Gene Aliases: |
MGN2, mago, magoh |
Location: |
Chr.12:10604190-10613609 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGOHB | NM_001300739.2 | 5 | 2371 | NP_001287668.1 |
| NM_001319985.1 | 5 | 2101 | NP_001306914.1 | |
| NM_018048.4 | 5 | 2203 | NP_060518.1 | |
| NR_135120.1 | 6 | 2489 | ||
| NR_135121.1 | 6 | 2387 | ||
| AK022720.1 | 5 | 2178 | BAB14202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832329 | Chr.12:10524495 - 10693347 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map