Assay Details
Target Gene Details
Entrez Gene ID: | 57458 |
Gene Name: | transmembrane and coiled-coil domain family 3 |
Gene Aliases: |
- |
Location: |
Chr.12:94567124-94650562 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMCC3 | NM_001301036.1 | 4 | 4585 | NP_001287965.1 |
| NM_020698.3 | 4 | 4566 | NP_065749.2 | |
| AB073655.1 | 1 | 3061 | ||
| BC040535.1 | 4 | 4552 | AAH40535.1 | |
| CA772279.1 | 1 | 640 |

Genomic Map