Assay Details
Target Gene Details
Entrez Gene ID: | 919 |
Gene Name: | CD247 molecule |
Gene Aliases: |
CD3-ZETA, CD3H, CD3Q, CD3Z, IMD25, T3Z, TCRZ |
Location: |
Chr.1:167430640-167518610 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD247 | NM_000734.3 | 8 | 708 | NP_000725.1 |
| NM_198053.2 | 8 | 711 | NP_932170.1 | |
| XM_011510144.2 | 9 | 779 | XP_011508446.1 | |
| XM_011510145.1 | 9 | 785 | XP_011508447.1 | |
| XM_017002800.1 | 9 | 770 | XP_016858289.1 | |
| XM_017002801.1 | 9 | 765 | XP_016858290.1 | |
| AK128376.1 | 7 | 2039 | BAC87407.1 | |
| AK223325.1 | 8 | 699 | BAD97045.1 | |
| AL557555.3 | ||||
| BC025703.1 | 8 | 627 | AAH25703.1 | |
| BI819937.1 | 8 | 685 | ||
| EF079882.1 | ||||
| EF364117.1 | ||||
| J04132.1 | 8 | 637 | AAA60394.1 | |
| X55510.1 | ||||
| X55511.1 | ||||
| X55512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890749 | Chr.1:167379314 - 167446554 on Build GRCh38 | Gain |
|
| nsv3432 | Chr.1:167418866 - 167452640 on Build GRCh38 | Insertion |
|
| nsv831814 | Chr.1:167380597 - 167590974 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map