Assay Details
Target Gene Details
Entrez Gene ID: | 3781 |
Gene Name: | potassium calcium-activated channel subfamily N member 2 |
Gene Aliases: |
KCa2.2, SK2, SKCA2, SKCa 2, hSK2 |
Location: |
Chr.5:114056006-114496500 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNN2 | NM_001278204.1 | 7 | 1893 | NP_001265133.1 |
| NM_021614.3 | 8 | 2312 | NP_067627.2 | |
| NM_170775.2 | 6 | 1254 | NP_740721.1 | |
| NR_103458.1 | 7 | 1360 | ||
| XM_011543387.1 | 13 | 3595 | XP_011541689.1 | |
| XM_011543389.1 | 12 | 3526 | XP_011541691.1 | |
| XM_017009457.1 | 6 | 1705 | XP_016864946.1 | |
| AF239613.1 | 8 | 2312 | AAG16728.1 | |
| AF397175.1 | AAK84039.1 | |||
| AK090521.1 | 1 | 1960 | ||
| AK289948.1 | 8 | 2244 | ||
| BC015371.2 | 6 | 1254 | AAH15371.1 | |
| BC117454.1 | ||||
| BC117456.1 | 8 | 2023 |
Target Gene Details
Entrez Gene ID: | 101927078 |
Gene Name: | uncharacterized LOC101927078 |
Gene Aliases: |
- |
Location: |
Chr.5:114447418-114773413 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927078 | NR_130785.1 | |||
| AK097686.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830452 | Chr.5:114462920 - 114639531 on Build GRCh38 | Loss |
|
| nsv599433 | Chr.5:114491590 - 114565393 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map