Assay Details
Target Gene Details
Entrez Gene ID: | 11096 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 5 |
Gene Aliases: |
ADAM-TS 11, ADAM-TS 5, ADAM-TS5, ADAMTS-11, ADAMTS-5, ADAMTS11, ADMP-2 |
Location: |
Chr.21:26917912-26967120 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS5 | NM_007038.4 | 8 | 5528 | NP_008969.2 |
| XM_011529448.2 | 7 | 5360 | XP_011527750.1 | |
| AF142099.1 | 8 | 4921 | AAD49577.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4420n100 | Chr.21:26910341 - 26924070 on Build GRCh38 | Loss |
|
| nsv834068 | Chr.21:26807351 - 26988489 on Build GRCh38 | Loss |
|
| nsv1066573 | Chr.21:26724037 - 27002035 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs143537582] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map