Assay Details
Target Gene Details
Entrez Gene ID: | 2051 |
Gene Name: | EPH receptor B6 |
Gene Aliases: |
HEP |
Location: |
Chr.7:142854640-142871093 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHB6 | NM_001280794.2 | NP_001267723.2 | ||
| NM_001280795.2 | NP_001267724.2 | |||
| NM_004445.5 | NP_004436.4 | |||
| NR_104001.2 | ||||
| XM_006715881.2 | XP_006715944.1 | |||
| XM_011515879.1 | XP_011514181.1 | |||
| XM_011515880.1 | XP_011514182.1 | |||
| XM_011515881.2 | XP_011514183.1 | |||
| XM_011515882.2 | XP_011514184.1 | |||
| BC110606.1 | AAI10607.1 | |||
| BC110607.1 | ||||
| BX410199.2 | ||||
| D83492.1 | BAA21560.1 | |||
| EU054308.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1020332 | Chr.7:142853182 - 142867988 on Build GRCh38 | Gain |
|
| nsv831168 | Chr.7:142830591 - 143025918 on Build GRCh38 | Gain |
|
| nsv949965 | Chr.7:142862343 - 142878755 on Build GRCh38 | Deletion |
|
| esv2759569 | Chr.7:142434958 - 143027060 on Build GRCh38 | Loss |
|
| nsv428187 | Chr.7:142618692 - 142909521 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map