Assay Details
Target Gene Details
Entrez Gene ID: | 158234 |
Gene Name: | tRNA methyltransferase 10B |
Gene Aliases: |
RG9MTD3, bA3J10.9 |
Location: |
Chr.9:37753204-37780459 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRMT10B | NM_001286950.1 | NP_001273879.1 | ||
| NM_001286951.1 | NP_001273880.1 | |||
| NM_001286952.1 | NP_001273881.1 | |||
| NM_001286953.1 | NP_001273882.1 | |||
| NM_001286954.1 | NP_001273883.1 | |||
| NM_144964.3 | NP_659401.2 | |||
| NR_104612.1 | ||||
| XM_005251375.2 | XP_005251432.1 | |||
| XM_005251379.2 | XP_005251436.1 | |||
| XM_011517735.2 | XP_011516037.1 | |||
| XM_011517736.2 | XP_011516038.1 | |||
| XM_011517738.2 | XP_011516040.1 | |||
| XM_011517739.2 | XP_011516041.1 | |||
| XM_011517742.1 | XP_011516044.1 | |||
| XM_011517743.2 | XP_011516045.1 | |||
| XM_017014313.1 | XP_016869802.1 | |||
| AK056017.1 | BAB71074.1 | |||
| AK092718.1 | BAC03957.1 | |||
| AK294219.1 | ||||
| AK295733.1 | ||||
| AK304516.1 | ||||
| AK315922.1 | ||||
| BC012175.1 | ||||
| BC057774.1 | AAH57774.1 | |||
| CR986760.1 | ||||
| HY066525.1 | 2 | 204 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469813 | Chr.9:37628200 - 37777281 on Build GRCh38 | Loss |
|
| esv3891658 | Chr.9:37689626 - 37790130 on Build GRCh38 | Gain |
|
| nsv831552 | Chr.9:37693652 - 37859980 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map