Assay Details
Target Gene Details
Entrez Gene ID: | 6873 |
Gene Name: | TATA-box binding protein associated factor 2 |
Gene Aliases: |
CIF150, MRT40, TAF2B, TAFII150 |
Location: |
Chr.8:119730774-119832858 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAF2 | NM_003184.3 | 26 | 4200 | NP_003175.1 |
| XM_006716621.3 | 27 | 4380 | XP_006716684.1 | |
| XM_011517259.2 | 26 | 4269 | XP_011515561.1 | |
| XM_017013779.1 | 25 | 4113 | XP_016869268.1 | |
| XM_017013780.1 | 26 | 3963 | XP_016869269.1 | |
| XM_017013781.1 | 27 | 4117 | XP_016869270.1 | |
| XM_017013782.1 | 25 | 4006 | XP_016869271.1 | |
| XM_017013783.1 | 26 | 3971 | XP_016869272.1 | |
| XM_017013784.1 | 26 | 3912 | XP_016869273.1 | |
| XM_017013785.1 | 27 | 4066 | XP_016869274.1 | |
| XM_017013786.1 | 25 | 4053 | XP_016869275.1 | |
| XM_017013787.1 | 25 | 3801 | XP_016869276.1 | |
| XM_017013788.1 | 25 | 3907 | XP_016869277.1 | |
| XM_017013790.1 | 24 | 3729 | XP_016869279.1 | |
| XM_017013791.1 | 27 | 4062 | XP_016869280.1 | |
| AK001618.1 | 11 | 1967 | ||
| AK001909.1 | 5 | 1151 | ||
| AL137601.1 | 1 | 160 | CAB70836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv612111 | Chr.8:119624610 - 119982154 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map