Assay Details
Target Gene Details
Entrez Gene ID: | 6996 |
Gene Name: | thymine DNA glycosylase |
Gene Aliases: |
hTDG |
Location: |
Chr.12:103965815-103988878 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TDG | NM_003211.4 | 1 | 87 | NP_003202.3 |
| XM_011538714.1 | XP_011537016.1 | |||
| XM_011538715.1 | XP_011537017.1 | |||
| AK299832.1 | ||||
| AK303631.1 | ||||
| AK309792.1 | ||||
| AK311673.1 | ||||
| AK313234.1 | ||||
| AL710869.1 | 1 | 81 | ||
| BC019925.1 | ||||
| BC037557.1 | AAH37557.1 | |||
| BC104477.1 | AAI04478.1 | |||
| CB047650.1 | ||||
| DB162106.1 | 1 | 87 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560039 | Chr.12:103783890 - 104061562 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map