Assay Details
Target Gene Details
Entrez Gene ID: | 3690 |
Gene Name: | integrin subunit beta 3 |
Gene Aliases: |
BDPLT16, BDPLT2, CD61, GP3A, GPIIIa, GT |
Location: |
Chr.17:47253842-47312711 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB3 | NM_000212.2 | 15 | 2401 | NP_000203.2 |
| BC127666.1 | 15 | 2401 | ||
| BC127667.1 | 15 | 2405 | ||
| J02703.1 | 15 | 2401 | AAA52589.1 | |
| KC120777.1 | 15 | 2402 | ||
| KC120778.1 | 15 | 2401 | ||
| M20311.1 | 15 | 2397 | AAA60122.1 | |
| M35999.1 | 15 | 2399 | AAA35927.1 | |
| U95204.1 | AAB71380.1 |
Target Gene Details
Entrez Gene ID: | 102724508 |
Gene Name: | thyroid cancer-associated transcript 158 |
Gene Aliases: |
- |
Location: |
Chr.17:47303443-47323901 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THCAT158 | NR_110880.1 | |||
| NR_110881.1 | ||||
| AA430297.1 | ||||
| AW293117.1 | ||||
| HY036509.1 |
More Information
Set Membership: |
|

Genomic Map