Assay Details
Target Gene Details
Entrez Gene ID: | 64321 |
Gene Name: | SRY-box 17 |
Gene Aliases: |
VUR3 |
Location: |
Chr.8:54457935-54460896 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOX17 | NM_022454.3 | NP_071899.1 | ||
| AK025905.1 | BAB15277.1 | |||
| BC030209.1 | ||||
| BC111365.1 | AAI11366.1 | |||
| BC111770.1 | AAI11771.1 | |||
| DA874945.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv438055 | Chr.8:54452024 - 54461327 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map