Assay Details
Target Gene Details
Entrez Gene ID: | 8659 |
Gene Name: | aldehyde dehydrogenase 4 family member A1 |
Gene Aliases: |
ALDH4, P5CD, P5CDh |
Location: |
Chr.1:18871430-18902799 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH4A1 | NM_001161504.1 | NP_001154976.1 | ||
| NM_001319218.1 | NP_001306147.1 | |||
| NM_003748.3 | NP_003739.2 | |||
| NM_170726.2 | NP_733844.1 | |||
| AK222486.1 | BAD96206.1 | |||
| AK289972.1 | ||||
| AK294552.1 | ||||
| BC007581.1 | AAH07581.1 | |||
| BC023600.2 | AAH23600.1 | |||
| FJ462711.1 | ||||
| U24266.1 | AAC50500.1 | |||
| U24267.1 | AAC50501.1 |
Target Gene Details
Entrez Gene ID: | 100616120 |
Gene Name: | microRNA 4695 |
Gene Aliases: |
- |
Location: |
Chr.1:18883202-18883275 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR4695 | NR_039844.2 |
More Information
Set Membership: |
|

Genomic Map