Assay Details
Target Gene Details
Entrez Gene ID: | 124446 |
Gene Name: | transmembrane protein 219 |
Gene Aliases: |
IGFBP-3R |
Location: |
Chr.16:29962030-29973052 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM219 | NM_001083613.1 | 4 | 572 | NP_001077082.1 |
| NM_194280.3 | 4 | 569 | NP_919256.1 | |
| XM_005255123.1 | 4 | 571 | XP_005255180.1 | |
| BC017488.1 | 3 | 337 | AAH17488.1 | |
| BC050051.1 | 4 | 599 | AAH50051.1 | |
| BF724791.1 | 4 | 529 | ||
| FJ748884.1 | 3 | 432 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3638377 | Chr.16:29967685 - 29971010 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79505862] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map