Assay Details
Target Gene Details
Entrez Gene ID: | 6184 |
Gene Name: | ribophorin I |
Gene Aliases: |
OST1, RBPH1 |
Location: |
Chr.3:128619970-128650876 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPN1 | NM_002950.3 | NP_002941.1 | ||
| AK223595.1 | BAD97315.1 | |||
| AK296905.1 | ||||
| AK297513.1 | ||||
| AK297941.1 | ||||
| AK312369.1 | ||||
| BC007995.1 | AAH07995.1 | |||
| BC010839.1 | AAH10839.1 | |||
| BG720812.1 | ||||
| CR456742.1 | CAG33023.1 | |||
| CR749284.1 | CAH18139.1 | |||
| Y00281.1 | CAA68392.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759176 | Chr.3:128338850 - 128718974 on Build GRCh38 | Loss |
|
| nsv508952 | Chr.3:128612661 - 128681787 on Build GRCh38 | Insertion |
|
| dgv4855n100 | Chr.3:128621905 - 128705613 on Build GRCh38 | Gain |
|
| esv2763764 | Chr.3:128637371 - 128695318 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map