Assay Details
Target Gene Details
Entrez Gene ID: | 344018 |
Gene Name: | folliculogenesis specific bHLH transcription factor |
Gene Aliases: |
BHLHC8, FIGALPHA, POF6 |
Location: |
Chr.2:70777310-70790643 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FIGLA | NM_001004311.3 | 3 | 504 | NP_001004311.2 |
| AY541030.1 | 3 | 504 | AAS48452.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv582192 | Chr.2:70752058 - 70797428 on Build GRCh38 | Loss |
|
| esv3892614 | Chr.2:70505720 - 70803944 on Build GRCh38 | Gain |
|
| esv2762674 | Chr.2:70752618 - 70829950 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map