Assay Details
Target Gene Details
Entrez Gene ID: | 8864 |
Gene Name: | period circadian clock 2 |
Gene Aliases: |
FASPS, FASPS1 |
Location: |
Chr.2:238244038-238290102 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PER2 | NM_022817.2 | 23 | 6249 | NP_073728.1 |
| XM_005246111.4 | 23 | 6235 | XP_005246168.1 | |
| XM_006712824.3 | 23 | 6187 | XP_006712887.1 | |
| AB002345.2 | 23 | 6134 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv584761 | Chr.2:238212011 - 238246412 on Build GRCh38 | Gain |
|
| nsv1000252 | Chr.2:235899999 - 238766417 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map