Assay Details
Target Gene Details
Entrez Gene ID: | 51280 |
Gene Name: | golgi membrane protein 1 |
Gene Aliases: |
C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3 |
Location: |
Chr.9:86026141-86100201 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLM1 | NM_016548.3 | 10 | 2087 | NP_057632.2 |
| NM_177937.2 | 10 | 2079 | NP_808800.1 | |
| AF236056.1 | 10 | 2028 | AAF44663.1 | |
| AJ420432.1 | 1 | 310 | ||
| AK027717.1 | 10 | 2050 | ||
| AK075542.1 | 10 | 2021 | BAC11685.1 | |
| AK075557.1 | 10 | 1991 | ||
| AK092943.1 | 11 | 1967 | ||
| BC001740.1 | 10 | 2002 | AAH01740.1 | |
| EU794616.1 | 10 | 2046 |
More Information
Additional Information:
For this assay, SNP(s) [rs114313531,rs142225824] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map