Assay Details
Target Gene Details
Entrez Gene ID: | 5210 |
Gene Name: | 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4 |
Gene Aliases: |
- |
Location: |
Chr.3:48517684-48562383 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PFKFB4 | NM_001317134.1 | 15 | 2265 | NP_001304063.1 |
| NM_001317135.1 | 14 | 2096 | NP_001304064.1 | |
| NM_001317136.1 | 14 | 2003 | NP_001304065.1 | |
| NM_001317137.1 | 13 | 2012 | NP_001304066.1 | |
| NM_001317138.1 | 15 | 2245 | NP_001304067.1 | |
| NM_004567.3 | 14 | 2117 | NP_004558.1 | |
| XM_011533829.2 | 14 | 2671 | XP_011532131.1 | |
| XM_017006614.1 | 15 | 2300 | XP_016862103.1 | |
| XM_017006615.1 | 14 | 2050 | XP_016862104.1 | |
| XM_017006616.1 | 14 | 1963 | XP_016862105.1 | |
| XM_017006617.1 | 14 | 2199 | XP_016862106.1 | |
| AF108765.1 | 14 | 1993 | AAD09427.1 | |
| BC010269.1 | 14 | 1988 | AAH10269.1 | |
| D49818.1 | BAA18921.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1004383 | Chr.3:48506361 - 48612339 on Build GRCh38 | Loss |
|
| nsv834684 | Chr.3:48344209 - 48551890 on Build GRCh38 | Gain |
|
| nsv523981 | Chr.3:48478880 - 48683607 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map