Assay Details
Target Gene Details
Entrez Gene ID: | 57626 |
Gene Name: | kelch like family member 1 |
Gene Aliases: |
MRP2 |
Location: |
Chr.13:69700593-70108493 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL1 | NM_001286725.1 | 8 | 2503 | NP_001273654.1 |
| NM_020866.2 | 9 | 2686 | NP_065917.1 | |
| XM_017020678.1 | 9 | 1606 | XP_016876167.1 | |
| XM_017020679.1 | 9 | 2193 | XP_016876168.1 | |
| AB040923.1 | 9 | 2652 | ||
| AF252283.1 | 9 | 2520 | AAF81719.1 | |
| AK291435.1 | 9 | 2686 | ||
| AK295939.1 | 8 | 1694 | ||
| BC022460.1 | 9 | 1930 | AAH22460.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045844 | Chr.13:69463964 - 69831303 on Build GRCh38 | Gain |
|
| nsv1049684 | Chr.13:69552739 - 69759313 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map