Assay Details
Target Gene Details
Entrez Gene ID: | 57818 |
Gene Name: | glucose-6-phosphatase catalytic subunit 2 |
Gene Aliases: |
IGRP |
Location: |
Chr.2:168901223-168910000 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| G6PC2 | NM_001081686.1 | 1 | 249 | NP_001075155.1 |
| NM_021176.2 | 1 | 249 | NP_066999.1 | |
| XM_011511564.2 | 1 | 266 | XP_011509866.1 | |
| XM_011511565.2 | XP_011509867.1 | |||
| BC104778.1 | 1 | 249 | AAI04779.1 | |
| BC113376.1 | 1 | 249 | ||
| BQ268961.1 | 1 | 184 | ||
| CR627438.1 | 1 | 200 | CAH10524.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583599 | Chr.2:168807717 - 169046586 on Build GRCh38 | Gain |
|
| nsv459943 | Chr.2:168900420 - 168942500 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map