Assay Details
Target Gene Details
Entrez Gene ID: | 1002 |
Gene Name: | cadherin 4 |
Gene Aliases: |
CAD4, R-CAD, RCAD |
Location: |
Chr.20:61252426-61940617 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDH4 | NM_001794.4 | NP_001785.2 | ||
| AK315192.1 | ||||
| BC101651.1 | AAI01652.1 | |||
| BC112150.1 | AAI12151.1 | |||
| L34059.1 | AAA35627.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7638n54 | Chr.20:61252058 - 61257735 on Build GRCh38 | Gain |
|
| nsv828767 | Chr.20:61247659 - 61259003 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map