Assay Details
Target Gene Details
Entrez Gene ID: | 23196 |
Gene Name: | family with sequence similarity 120A |
Gene Aliases: |
C9orf10, OSSA |
Location: |
Chr.9:93451696-93566115 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM120A | NM_001286722.1 | NP_001273651.1 | ||
| NM_001286723.1 | NP_001273652.1 | |||
| NM_001286724.1 | NP_001273653.1 | |||
| NM_014612.4 | NP_055427.2 | |||
| XM_005251842.4 | XP_005251899.1 | |||
| XM_011518412.2 | XP_011516714.1 | |||
| XM_011518413.2 | XP_011516715.1 | |||
| XM_011518414.2 | XP_011516716.1 | |||
| XM_011518416.2 | XP_011516718.1 | |||
| AF214737.1 | AAF72866.1 | |||
| AF214738.1 | AAF72867.1 | |||
| BC098584.1 | AAH98584.1 | |||
| BC111736.1 | AAI11737.1 | |||
| D80005.2 | BAA11500.2 |
Target Gene Details
Entrez Gene ID: | 158293 |
Gene Name: | family with sequence similarity 120A opposite strand |
Gene Aliases: |
C9orf10OS |
Location: |
Chr.9:93446500-93453592 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM120AOS | NM_198841.2 | NP_942138.2 | ||
| NR_136229.1 | ||||
| NR_136230.1 | ||||
| NR_136231.1 | ||||
| AK056096.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950899 | Chr.9:93453119 - 93478818 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map