Assay Details
Target Gene Details
Entrez Gene ID: | 102465507 |
Gene Name: | microRNA 6842 |
Gene Aliases: |
hsa-mir-6842 |
Location: |
Chr.8:27433370-27433434 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6842 | NR_106901.1 |
Target Gene Details
Entrez Gene ID: | 2185 |
Gene Name: | protein tyrosine kinase 2 beta |
Gene Aliases: |
CADTK, CAKB, FADK2, FAK2, PKB, PTK, PYK2, RAFTK |
Location: |
Chr.8:27311482-27459391 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTK2B | NM_004103.4 | NP_004094.3 | ||
| NM_173174.2 | NP_775266.1 | |||
| NM_173175.2 | NP_775267.1 | |||
| NM_173176.2 | NP_775268.1 | |||
| XM_005273447.4 | XP_005273504.1 | |||
| XM_005273448.4 | XP_005273505.1 | |||
| XM_011544441.2 | XP_011542743.1 | |||
| XM_011544442.2 | XP_011542744.1 | |||
| XM_017013214.1 | XP_016868703.1 | |||
| XM_017013215.1 | XP_016868704.1 | |||
| XM_017013216.1 | XP_016868705.1 | |||
| AB209085.1 | BAD92322.1 | |||
| AK128371.1 | BAC87404.1 | |||
| AY429564.1 | ||||
| BC036651.2 | AAH36651.1 | |||
| BC042599.2 | AAH42599.1 | |||
| L49207.1 | AAB47217.1 | |||
| U33284.1 | AAC50203.1 | |||
| U43522.1 | AAC05330.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3616721 | Chr.8:27375902 - 27443884 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map