Assay Details
Target Gene Details
Entrez Gene ID: | 4738 |
Gene Name: | neural precursor cell expressed, developmentally down-regulated 8 |
Gene Aliases: |
NEDD-8 |
Location: |
Chr.14:24216851-24232370 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEDD8 | NM_006156.2 | NP_006147.1 | ||
| BC104200.1 | AAI04201.1 | |||
| BC104201.1 | AAI04202.1 | |||
| BC104664.1 | AAI04665.1 | |||
| BE548741.1 | ||||
| CF130271.1 | ||||
| CR407662.1 | CAG28590.1 | |||
| D23662.1 | BAA04889.1 |
More Information
Set Membership: |
|

Genomic Map