Assay Details
Target Gene Details
Entrez Gene ID: | 51063 |
Gene Name: | calcium homeostasis modulator 2 |
Gene Aliases: |
FAM26B |
Location: |
Chr.10:103446785-103452419 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALHM2 | NM_015916.4 | 3 | 626 | NP_057000.2 |
| NR_024552.1 | 3 | 523 | ||
| NR_046344.1 | 3 | 523 | ||
| XM_006717880.2 | 3 | 502 | XP_006717943.1 | |
| XM_006717883.2 | 2 | 387 | XP_006717946.1 | |
| XM_006717884.3 | 3 | 1146 | XP_006717947.1 | |
| XM_011539848.2 | 3 | 1158 | XP_011538150.1 | |
| XM_017016306.1 | 3 | 1085 | XP_016871795.1 | |
| XM_017016307.1 | 3 | 992 | XP_016871796.1 | |
| XM_017016308.1 | 3 | 640 | XP_016871797.1 | |
| XM_017016309.1 | 2 | 387 | XP_016871798.1 | |
| AF131810.1 | 3 | 384 | AAD20050.1 | |
| AK022195.1 | 3 | 521 | BAB13983.1 | |
| AK125276.1 | 3 | 1213 | ||
| BC000039.2 | 3 | 539 | AAH00039.1 | |
| DB109582.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524966 | Chr.10:103441820 - 103717405 on Build GRCh38 | Loss |
|
| nsv518987 | Chr.10:103434329 - 103484808 on Build GRCh38 | Loss |
|
| dgv966n100 | Chr.10:103385173 - 103553473 on Build GRCh38 | Gain |
|
| dgv280e212 | Chr.10:103390805 - 103464349 on Build GRCh38 | Gain |
|
| esv3577694 | Chr.10:103391276 - 103553473 on Build GRCh38 | Gain |
|
| esv3624414 | Chr.10:103412843 - 103459270 on Build GRCh38 | Gain |
|
| nsv552060 | Chr.10:103447748 - 103470575 on Build GRCh38 | Loss |
|
| esv3624413 | Chr.10:103392636 - 103459758 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map