Assay Details
Target Gene Details
Entrez Gene ID: | 114899 |
Gene Name: | C1q and tumor necrosis factor related protein 3 |
Gene Aliases: |
C1ATNF3, CORCS, CORS, CORS-26, CORS26, CTRP3 |
Location: |
Chr.5:34017858-34043266 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1QTNF3 | NM_030945.3 | 4 | 550 | NP_112207.1 |
| NM_181435.5 | 4 | 769 | NP_852100.3 | |
| AF173888.1 | 4 | 452 | AAQ13635.1 | |
| AF326976.1 | 4 | 485 | AAK70344.1 | |
| AF329837.1 | 4 | 492 | AAK17961.1 | |
| AK075533.1 | 4 | 497 | BAC11676.1 | |
| AK295968.1 | 4 | 769 | ||
| AK315921.1 | 4 | 756 | ||
| AY358388.1 | 4 | 485 | AAQ88754.1 | |
| BC112925.1 | 4 | 424 | ||
| BC120990.2 | 4 | 699 | ||
| BX640995.1 | 5 | 372 | CAE45998.1 | |
| EU399231.1 | 4 | 414 | ||
| EU399232.1 | 4 | 633 |
Target Gene Details
Entrez Gene ID: | 100534612 |
Gene Name: | C1QTNF3-AMACR readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.5:33986986-34124528 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1QTNF3-AMACR | NR_037951.1 | 4 | 436 | |
| AB209261.1 | 4 | 445 | BAD92498.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830255 | Chr.5:33929507 - 34109160 on Build GRCh38 | Loss |
|
| nsv519240 | Chr.5:33950598 - 34749311 on Build GRCh38 | Gain |
|
| nsv522715 | Chr.5:33319157 - 34037875 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map