Assay Details
Target Gene Details
Entrez Gene ID: | 51686 |
Gene Name: | ornithine decarboxylase antizyme 3 |
Gene Aliases: |
AZ3, OAZ-t, TISP15 |
Location: |
Chr.1:151762969-151771330 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OAZ3 | NM_001134939.1 | NP_001128411.1 | ||
| NM_001301371.1 | NP_001288300.1 | |||
| NM_016178.2 | NP_057262.2 | |||
| AF175296.1 | AAD51734.1 | |||
| AK125876.1 | ||||
| AY713300.1 | ||||
| BC073949.1 | AAH73949.1 | |||
| BI830988.1 |
Target Gene Details
Entrez Gene ID: | 11022 |
Gene Name: | tudor and KH domain containing |
Gene Aliases: |
TDRD2 |
Location: |
Chr.1:151769354-151790576 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TDRKH | XM_017000123.1 | XP_016855612.1 | ||
| AK225160.1 |
More Information
Set Membership: |
|

Genomic Map