Assay Details
Target Gene Details
Entrez Gene ID: | 3687 |
Gene Name: | integrin subunit alpha X |
Gene Aliases: |
CD11C, SLEB6 |
Location: |
Chr.16:31355134-31382997 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGAX | NM_000887.4 | NP_000878.2 | ||
| NM_001286375.1 | NP_001273304.1 | |||
| XM_011545852.1 | XP_011544154.1 | |||
| XM_011545854.1 | XP_011544156.1 | |||
| AK296666.1 | ||||
| AK309642.1 | ||||
| BC038237.1 | AAH38237.1 | |||
| BM149283.1 | ||||
| DA324567.1 | ||||
| M81695.1 | AAA59180.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833194 | Chr.16:31346858 - 31517224 on Build GRCh38 | Loss |
|
| nsv1061980 | Chr.16:31327141 - 31404150 on Build GRCh38 | Gain |
|
| dgv503e214 | Chr.16:31332171 - 31413104 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map