Assay Details
Target Gene Details
Entrez Gene ID: | 6760 |
Gene Name: | SS18, nBAF chromatin remodeling complex subunit |
Gene Aliases: |
SSXT, SYT |
Location: |
Chr.18:26016253-26091217 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SS18 | NM_001007559.2 | 11 | 1360 | NP_001007560.1 |
| NM_001308201.1 | 11 | 1335 | NP_001295130.1 | |
| NM_005637.3 | 10 | 1267 | NP_005628.2 | |
| XM_006722527.2 | 12 | 1386 | XP_006722590.1 | |
| XM_011526145.1 | 11 | 1328 | XP_011524447.1 | |
| XM_011526147.2 | 13 | 1611 | XP_011524449.1 | |
| XM_011526148.2 | 13 | 1598 | XP_011524450.1 | |
| XM_011526149.2 | 13 | 1623 | XP_011524451.1 | |
| XM_011526150.2 | 12 | 1483 | XP_011524452.1 | |
| XM_011526151.2 | 13 | 1612 | XP_011524453.1 | |
| XM_011526152.2 | 11 | 1390 | XP_011524454.1 | |
| AF343880.1 | 11 | 1294 | AAK21314.1 | |
| AK289510.1 | 11 | 1338 | ||
| AK296949.1 | 11 | 1318 | ||
| AK299082.1 | 10 | 1228 | ||
| AK301504.1 | 10 | 1272 | ||
| AK304305.1 | 11 | 1383 | ||
| AK316302.1 | 10 | 1217 | ||
| AK316338.1 | 11 | 1336 | ||
| BC096221.1 | AAH96221.1 | |||
| BC096222.1 | AAH96222.1 | |||
| BC096223.1 | AAH96223.1 | |||
| BC096224.1 | AAH96224.1 | |||
| CR542088.1 | CAG46885.1 | |||
| CR542103.1 | CAG46900.1 | |||
| X79201.1 | 10 | 1204 | CAA55792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893078 | Chr.18:25391998 - 26039471 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map