Assay Details
Target Gene Details
Entrez Gene ID: | 23644 |
Gene Name: | enhancer of mRNA decapping 4 |
Gene Aliases: |
GE1, Ge-1, HEDL5, HEDLS, RCD-8, RCD8 |
Location: |
Chr.16:67873023-67884514 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EDC4 | NM_014329.4 | NP_055144.3 | ||
| AK128582.1 | ||||
| AK291049.1 | ||||
| AK302563.1 | ||||
| BC043616.1 | AAH43616.1 | |||
| BC053598.1 | AAH53598.1 | |||
| BC064567.1 | AAH64567.1 | |||
| L26339.1 | AAA21833.1 | |||
| U17474.1 | AAB51444.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522852 | Chr.16:67153308 - 68071571 on Build GRCh38 | Gain |
|
| nsv1844 | Chr.16:67868325 - 67917356 on Build GRCh38 | Deletion |
|
| nsv524363 | Chr.16:67136529 - 68071571 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map