Assay Details
Target Gene Details
Entrez Gene ID: | 4956 |
Gene Name: | outer dense fiber of sperm tails 1 |
Gene Aliases: |
CT133, HSPB10, ODF, ODF2, ODF27, ODFP, ODFPG, ODFPGA, ODFPGB, RT7, SODF |
Location: |
Chr.8:102551620-102561017 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ODF1 | NM_024410.3 | 1 | 201 | NP_077721.2 |
| AK315276.1 | 1 | 194 | ||
| BC104456.1 | 1 | 172 | AAI04457.1 | |
| BC104457.1 | 1 | 172 | AAI04458.1 | |
| DB457574.1 | 1 | 201 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576598 | Chr.8:102200959 - 102574604 on Build GRCh38 | Gain |
|
| esv2762761 | Chr.8:102480128 - 102613016 on Build GRCh38 | Gain |
|
| esv3618224 | Chr.8:102532605 - 102572593 on Build GRCh38 | Gain |
|
| esv33118 | Chr.8:102346339 - 103055944 on Build GRCh38 | Loss |
|
| nsv1024876 | Chr.8:102500557 - 102564430 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map