Assay Details
Target Gene Details
Entrez Gene ID: | 81631 |
Gene Name: | microtubule associated protein 1 light chain 3 beta |
Gene Aliases: |
ATG8F, LC3B, MAP1A/1BLC3, MAP1LC3B-a |
Location: |
Chr.16:87392195-87404774 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP1LC3B | NM_022818.4 | 1 | 107 | NP_073729.1 |
| AF087871.1 | AAM10499.1 | |||
| AF183417.1 | AAG09686.1 | |||
| AK025556.1 | BAB15169.1 | |||
| AK290956.1 | ||||
| AK310224.1 | ||||
| AK310335.1 | ||||
| BC041874.1 | 1 | 502 | AAH41874.1 | |
| BC067797.1 | AAH67797.1 | |||
| BG771621.1 | 1 | 92 | ||
| DA577457.1 | ||||
| DB055844.1 | 1 | 107 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv499n67 | Chr.16:87353978 - 87409229 on Build GRCh38 | Gain |
|
| nsv1131120 | Chr.16:87392294 - 87392794 on Build GRCh38 | Deletion |
|
| nsv833320 | Chr.16:87315844 - 87501226 on Build GRCh38 | Gain |
|
| nsv510694 | Chr.16:87276349 - 87400084 on Build GRCh38 | Deletion |
|
| nsv428330 | Chr.16:87336922 - 87633082 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs145549507,rs74605057] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map