Assay Details
Target Gene Details
Entrez Gene ID: | 392 |
Gene Name: | Rho GTPase activating protein 1 |
Gene Aliases: |
CDC42GAP, RHOGAP, RHOGAP1, p50rhoGAP |
Location: |
Chr.11:46677075-46700665 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP1 | NM_004308.3 | NP_004299.1 | ||
| XM_011520095.1 | XP_011518397.1 | |||
| XM_017017720.1 | XP_016873209.1 | |||
| AK126404.1 | ||||
| AK298562.1 | ||||
| AK307296.1 | ||||
| AK310605.1 | 3 | 325 | ||
| BC018118.1 | AAH18118.1 | |||
| HY004921.1 | ||||
| U02570.1 | AAA16142.1 | |||
| Z23024.1 | CAA80560.1 |
More Information
Set Membership: |
|

Genomic Map