Assay Details
Target Gene Details
Entrez Gene ID: | 1843 |
Gene Name: | dual specificity phosphatase 1 |
Gene Aliases: |
CL100, HVH1, MKP-1, MKP1, PTPN10 |
Location: |
Chr.5:172768090-172771200 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUSP1 | NM_004417.3 | 4 | 1391 | NP_004408.1 |
| AK298047.1 | 5 | 1325 | ||
| AK298658.1 | 5 | 1191 | ||
| AK299391.1 | ||||
| AK300487.1 | 5 | 1305 | ||
| AK304475.1 | 5 | 1183 | ||
| AK316321.1 | 5 | 1134 | ||
| BC022463.1 | 4 | 1386 | AAH22463.1 | |
| X68277.1 | 4 | 1376 | CAA48338.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600294 | Chr.5:172768130 - 172771179 on Build GRCh38 | Loss |
|
| esv2759393 | Chr.5:172554329 - 172788409 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map