Assay Details
Target Gene Details
Entrez Gene ID: | 339416 |
Gene Name: | ankyrin repeat domain 45 |
Gene Aliases: |
CT117 |
Location: |
Chr.1:173608336-173705430 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKRD45 | NM_198493.2 | 6 | 1056 | NP_940895.1 |
| XM_011509466.2 | 7 | 1503 | XP_011507768.1 | |
| XM_011509467.1 | 8 | 2027 | XP_011507769.1 | |
| XM_011509468.1 | 7 | 1995 | XP_011507770.1 | |
| XM_011509470.1 | 6 | 1711 | XP_011507772.1 | |
| XM_017001123.1 | 8 | 1365 | XP_016856612.1 | |
| AK127170.1 | 6 | 1056 | BAC86865.1 | |
| BC126353.1 | ||||
| BC126355.1 |
Target Gene Details
Entrez Gene ID: | 730159 |
Gene Name: | uncharacterized LOC730159 |
Gene Aliases: |
- |
Location: |
Chr.1:173603003-173637134 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC730159 | XM_011509968.2 | XP_011508270.1 |
More Information
Set Membership: |
|

Genomic Map