Assay Details
Target Gene Details
Entrez Gene ID: | 2 |
Gene Name: | alpha-2-macroglobulin |
Gene Aliases: |
A2MD, CPAMD5, FWP007, S863-7 |
Location: |
Chr.12:9067708-9115962 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A2M | NM_000014.4 | 30 | 3938 | NP_000005.2 |
| XM_006719056.2 | 30 | 3916 | XP_006719119.1 | |
| AB209614.2 | 30 | 3849 | BAD92851.1 | |
| AK307832.1 | 29 | 3675 | ||
| AK312482.1 | 30 | 3838 | ||
| AY591530.1 | 30 | 3903 | AAT02228.1 | |
| BC026246.1 | AAH26246.1 | |||
| BC040071.1 | 30 | 3877 | AAH40071.1 | |
| BX647329.1 | 13 | 1615 | ||
| CR749334.1 | 30 | 3896 | CAH18188.1 | |
| M11313.1 | 30 | 3868 | AAA51551.1 | |
| M36501.1 | 11 | 1332 | AAA51552.1 |
Target Gene Details
Entrez Gene ID: | 10219 |
Gene Name: | killer cell lectin like receptor G1 |
Gene Aliases: |
2F1, CLEC15A, MAFA, MAFA-2F1, MAFA-L, MAFA-LIKE |
Location: |
Chr.12:8989079-9255692 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLRG1 | XM_017018683.1 | XP_016874172.1 | ||
| XM_017018684.1 | XP_016874173.1 | |||
| XM_017018685.1 | XP_016874174.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
| nsv832327 | Chr.12:8929183 - 9093624 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map