Assay Details
Target Gene Details
Entrez Gene ID: | 8111 |
Gene Name: | G protein-coupled receptor 68 |
Gene Aliases: |
GPR12A, OGR1 |
Location: |
Chr.14:91232532-91264581 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR68 | NM_001177676.1 | 2 | 642 | NP_001171147.1 |
| NM_003485.3 | 2 | 596 | NP_003476.3 | |
| XM_005268110.4 | 2 | 1782 | XP_005268167.1 | |
| XM_005268111.3 | 2 | 562 | XP_005268168.1 | |
| XM_005268112.3 | 2 | 502 | XP_005268169.1 | |
| XM_006720262.3 | 2 | 528 | XP_006720325.1 | |
| XM_011537196.2 | 3 | 646 | XP_011535498.1 | |
| XM_011537197.2 | 2 | 926 | XP_011535499.1 | |
| XM_011537198.2 | 2 | 1152 | XP_011535500.1 | |
| XM_011537199.2 | 2 | 597 | XP_011535501.1 | |
| AY615372.1 | 1 | 331 | AAT38917.1 | |
| BC067470.1 | 1 | 301 | AAH67470.1 | |
| BC067471.1 | 1 | 301 | AAH67471.1 | |
| BC067472.1 | 1 | 301 | AAH67472.1 | |
| BC069592.1 | 1 | 301 | AAH69592.1 | |
| BC096071.3 | 1 | 367 | AAH96071.1 | |
| BC096072.3 | 1 | 367 | AAH96072.1 | |
| BC096073.1 | 1 | 367 | AAH96073.1 | |
| BC096074.3 | 1 | 367 | AAH96074.1 | |
| BC098567.1 | 2 | 539 | AAH98567.1 | |
| BC128536.1 | 2 | 650 | ||
| BM723705.1 | 2 | 641 | ||
| EU883575.1 | 1 | 331 | ||
| U35398.1 | 1 | 401 | AAA79060.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046924 | Chr.14:90947458 - 91371146 on Build GRCh38 | Gain |
|
| nsv832857 | Chr.14:91185830 - 91325925 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map