Assay Details
Target Gene Details
Entrez Gene ID: | 9108 |
Gene Name: | myotubularin related protein 7 |
Gene Aliases: |
- |
Location: |
Chr.8:17296794-17413531 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTMR7 | NM_004686.4 | 14 | 2218 | NP_004677.3 |
| XM_006716414.3 | 9 | 2584 | XP_006716477.2 | |
| XM_011544692.2 | 11 | 1657 | XP_011542994.1 | |
| XM_011544693.2 | 10 | 2119 | XP_011542995.1 | |
| XM_017013956.1 | 13 | 1907 | XP_016869445.1 | |
| AK294468.1 | 14 | 1979 | ||
| AK303762.1 | 6 | 1041 | ||
| AK303803.1 | 5 | 991 | ||
| AK316022.1 | 14 | 2028 | ||
| AK316283.1 | 14 | 2038 | ||
| BC130577.1 | 14 | 2004 | ||
| BC130579.1 | 14 | 2004 | ||
| CR749240.1 | 14 | 2016 | CAH18096.1 |
Target Gene Details
Entrez Gene ID: | 137492 |
Gene Name: | VPS37A, ESCRT-I subunit |
Gene Aliases: |
HCRP1, PQBP2, SPG53 |
Location: |
Chr.8:17246892-17333532 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS37A | XM_017013022.1 | XP_016868511.1 | ||
| XM_017013025.1 | XP_016868514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831245 | Chr.8:17255574 - 17450899 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map