Assay Details
Target Gene Details
Entrez Gene ID: | 10144 |
Gene Name: | family with sequence similarity 13 member A |
Gene Aliases: |
ARHGAP48, FAM13A1 |
Location: |
Chr.4:88725954-89111398 on Build GRCh38 |
Assay Gene Location: | Within Exon 31 |
Target Gene Details
Entrez Gene ID: | 285512 |
Gene Name: | FAM13A antisense RNA 1 |
Gene Aliases: |
FAM13A1OS, FAM13AOS, NCRNA00039 |
Location: |
Chr.4:88709789-88730103 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM13A-AS1 | NR_002806.2 | |||
| AK093650.1 | ||||
| AK126217.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470051 | Chr.4:88723780 - 89721993 on Build GRCh38 | Gain |
|
| nsv461576 | Chr.4:88621130 - 90539537 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74629576] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map