Assay Details
Target Gene Details
Entrez Gene ID: | 79918 |
Gene Name: | SET domain containing 6 |
Gene Aliases: |
- |
Location: |
Chr.16:58515479-58521961 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETD6 | NM_001160305.2 | 8 | 1618 | NP_001153777.1 |
| NM_024860.3 | 9 | 1546 | NP_079136.2 | |
| NR_134583.1 | 9 | 1577 | ||
| AK024801.1 | 9 | 1502 | BAB15011.1 | |
| AK127136.1 | 5 | 3023 | BAC86845.1 |
More Information
Set Membership: |
|

Genomic Map