Assay Details
Target Gene Details
Entrez Gene ID: | 9572 |
Gene Name: | nuclear receptor subfamily 1 group D member 1 |
Gene Aliases: |
EAR1, THRA1, THRAL, ear-1, hRev |
Location: |
Chr.17:40092784-40100725 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR1D1 | NM_021724.4 | NP_068370.1 | ||
| AK300155.1 | ||||
| BC047875.1 | ||||
| BC056148.1 | AAH56148.1 | |||
| BM671861.1 | ||||
| M24898.1 | AAA52335.1 | |||
| X72631.1 | CAB53540.1 |
Target Gene Details
Entrez Gene ID: | 7067 |
Gene Name: | thyroid hormone receptor, alpha |
Gene Aliases: |
AR7, CHNG6, EAR7, ERB-T-1, ERBA, ERBA1, NR1A1, THRA1, THRA2, c-ERBA-1 |
Location: |
Chr.17:40062193-40093867 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THRA | NM_001190918.1 | NP_001177847.1 | ||
| NM_001190919.1 | NP_001177848.1 | |||
| NM_003250.5 | NP_003241.2 | |||
| AF522368.1 | AAM77692.1 | |||
| AK290530.1 | ||||
| BC000261.1 | AAH00261.1 | |||
| BC002728.2 | AAH02728.1 | |||
| BC035137.1 | AAH35137.1 | |||
| J03239.1 | AAA61176.1 | |||
| M24899.1 | AAA35783.1 | |||
| M24900.1 | 1 | 1475 | AAA52333.1 | |
| Y00479.1 | CAA68539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833444 | Chr.17:40086570 - 40281028 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map